Thermo Scientific™ anti-Interleukin 1 beta Recombinant, Thermo Scientific™

Interleukin 1 beta Recombinant protein SIL1B from Thermo Scientific™ specifically detects Interleukin 1 beta in Not Applicable samples, and is validated for Control, ELISA and WB

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: EH2IL1B2

HUMAN IL-1B ELISA

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10648094

Quantity Price
1 £ 600.0 / 1 set
EU Stock 3
Add to basket


Description and Specification

Specification

Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sensitivity ≤ 1pg/mL
Formulation Complete ELISA kit with coated 96-well plates
Measurement Absorbance at 450nm minus absorbance at 550nm
Kit Contents 30X Wash Buffer, 50mL
Assay Range 10.2 to 400pg/mL
Sufficient For 2 x 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Specificity No significant cross-reaction with human IL-1a, IL-1RA, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, IFNa, IFNg or mouse IL-1ß
Sample Size 50µL
Target Natural and recombinant human interleukin-1 beta (IL-1ß)
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Description Human IL-1 beta ELISA Kit
Total Assay Time 4 hr.
Quantity 2 plate kit

These human IL-1 beta ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-1 beta (IL-1β) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-1β antibody pairs and IL-1β standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-1 beta (IL-1β)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 10.2 to 400pg/mL

  • Sensitivity: ≤ 1pg/mL

  • Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1RA, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, IFNα, IFNγ or mouse IL-1β

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).