missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems™

Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SLA50

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452796

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-1 alpha/IL-1F1 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).
Specifications

Specifications

1/Pk.
Human
HRP
ELISA
Solid Phase Sandwich ELISA
1pg/mL
Natural and recombinant human IL-1 alpha
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
3.90 - 250pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3552
6 Plates
Cell Culture Supernates (200uL),Serum (200uL),EDTA Plasma (200uL),Heparin Plasma (200uL),Citrate Plasma (200uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Videos
Documents