missing translation for 'onlineSavingsMsg'
Learn More

IgM Mouse anti-Human, Super Bright 436, Clone: SA-DA4, eBioscience™

Mouse Monoclonal Antibody

£118.00 - £249.00

Specifications

Antigen IgM
Clone SA-DA4
Target Species Human
Conjugate Super Bright 436
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15845543
View Documents
Invitrogen
62-9998-41
25 Tests
£118.00
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
15855543
View Documents
Invitrogen
62-9998-42
100 Tests
£249.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

IgM
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
SA-DA4
Super Bright 436
Flow Cytometry
Primary
Monoclonal
PBS with BSA and 0.09% sodium azide; pH 7.2
RUO
SDS