Learn More
Invitrogen™ eBioscience™ Human IgM Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human IgM.
Brand:  Invitrogen™ BMS2098FIVECE
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15561647
  • £1242.12 / Pack of 5
Estimated Shipment date
from Supplier 26-02-2018
Add to basket
Disclaimers:
For Research Use Only
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgM
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
HRP
5 x 96 tests
Serum, plasma
Human
Immunoglobulin M
ELISA
1ng/mL
50μL (1:10,000 prediluted)
IgM
Videos