Learn More
Invitrogen™ eBioscience™ Human IgM Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human IgM.
Brand:  Invitrogen™ BMS2098TWOCE
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15591647
  • £520.00 / Pack of 2
Estimated Shipment date
from Supplier 06-03-2020
Add to basket
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgM
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
HRP
1ng/mL
IgM
2 x 96 Tests
Serum
ELISA
50 μL (1:10,000 prediluted)
Immunoglobulin M