Learn More
Invitrogen™ IgG (Total) Human Uncoated ELISA Kit with Plates
For quantitative detection of human total IgG.
£188.00 - £906.00
Specifications
Assay Range | 1.6-100 ng/mL |
---|---|
Assay Sensitivity | 1.6 ng/mL |
Conjugate | HRP |
Product Type | ELISA Kit |
Sample Type | Plasma, Serum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15580937
|
Invitrogen™
88-50550-76 |
20 x 96 Tests |
£906.00
Pack of 20 |
Estimated Shipment: 27-03-2024
Log in to see stock. |
|||||
15500947
|
Invitrogen™
88-50550-86 |
10 x 96 Tests |
£452.00
Pack of 10 |
Estimated Shipment: 27-03-2024
Log in to see stock. |
|||||
15570937
|
Invitrogen™
88-50550-22 |
2 x 96 Tests |
£188.00
Pack of 2 |
Estimated Shipment: 25-03-2024
Log in to see stock. |
|||||
Description
The Human Total Immunoglobulin G (igG) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human igG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human IgG total Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Spezifikation
1.6-100 ng/mL | |
HRP | |
Plasma, Serum | |
ELISA | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
1.6 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
HRP | |
Plasma, 10 μL; Serum, 10 μL | |
Immunoglobulin G | |
1 hr. 45 min. |
For Research Use Only