missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IGFBP-3 Quantikine ELISA Kit, R&D Systems™

Human IGFBP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DGB300

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432655

  • £621.81 / 96 tests


Estimated Shipment date
from Supplier 01-04-2020
Add to basket

Description

Description

The Human IGFBP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IGFBP-3 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.14ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Serum
0.78 - 50ng/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
3486
1 Plate
Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents