missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IGFBP-1 DuoSet, R&D Systems™

Human IGFBP-1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DY871

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454155

  • £715.89 / Pack of 15


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

The Human IGFBP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IGFBP-1.
Specifications

Specifications

ELISA
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
15 96-well plates
Biotin
3484
Human
31.20 to 2000 pg/mL
Videos
Documents