missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IGFBP-1 DuoSet, R&D Systems™
Human IGFBP-1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 31.20 to 2000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 3484 |
Product Type | ELISA Kit |
Format | 15 96-well plates |
Description
The Human IGFBP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IGFBP-1.Specifications
31.20 to 2000 pg/mL | |
3484 | |
15 96-well plates | |
Human |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Kit | |
Biotin |