missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IgE Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgE.

Brand:  Invitrogen™ BMS2097TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531647

  • £2174.80 / Pack of 10


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgE

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
10 x 96 tests
Serum, plasma
Human
IgE
ELISA
0.5ng/mL
10μL
2° to 8°C
Immunoglobulin E
Videos

For Research Use Only