missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IgA Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IgA.

Brand:  Invitrogen™ BMS2096CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571637

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgA

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Serum, plasma
Human
IgA
ELISA
0.03ng/mL
20μL (1:10,000 prediluted)
2° to 8°C
Immunoglobulin A
Videos

For Research Use Only