missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IFN-omega Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IFN-omega.

Brand:  Invitrogen™ BMS233TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551787

  • £2175.09 / Pack of 10


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-omega
Biotin-Conjugate anti-human IFN-omega monoclonal antibody
Streptavidin-HRP
Human IFN-omega Standard, 6000pg/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
IFNWP2, Interferon alpha-II-1, Interferon omega-1
ELISA
1.5pg/mL
25μL
IFN omega
Videos

For Research Use Only