Learn More
Invitrogen™ Human IFN-gamma Recombinant Protein
Human IFN-gamma Recombinant Protein
Brand: Invitrogen™ RIFNG100
Code : PN
Additional Details : CAS Number : 5675-51-4 Weight : 0.80000kg
Description
Human IFN-gamma Recombinant Protein for Western Blot, IP, ELISA
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia. For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: One unit of activity is <0.05ng/mL, corresponding to a specific activity of 2x10^7 units/mg, as determined in a virus resistance assay. Reconstitute with 0.5 mL of distilled water.Specifications
P01579 | |
PBS with no preservative | |
Human IFN gamma | |
E. Coli | |
RUO | |
Human IFN gamma | |
Human | |
Hu | |
Recombinant | |
Recombinant |
ELISA, Immunoprecipitation, Western Blot | |
3458 | |
100μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human Interferon Gamma IFNG | |
IFNG | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |