missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IFN-gamma Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IFN-gamma.

Brand:  Invitrogen™ BMS228CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15541767

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-gamma
Biotin-Conjugate anti-human IFN-gamma monoclonal antibody
Streptavidin-HRP
Human IFN-gamma Standard lyophilized, 200ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Immune interferon, IFG
ELISA
0.99pg/mL
50μL
IFN gamma
Videos

For Research Use Only