missing translation for 'onlineSavingsMsg'
Learn More

eBioscience™ Human IFN gamma ELISPOT ELISA Ready-SET-Go!™ Kit

This ELISPOT reagent set is pre-titrated for optimal spot development.

Brand:  Affymetrix eBioscience 88-7386-21

UNSPSC : 12000000

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12730170

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating BufferThis Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
5X ELISA/ELISPOT Diluent.
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Specifications

Description

Human IFN gamma ELISPOT Ready-SET-Go! reagent set contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IFN gamma-secreting cells.
Description & Specifications

Specifications

ELISPOT
Human
IFN gamma
2 x 96 tests
2° to 8°C
Interferon gamma