missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-beta ELISA Kit, R&D Systems™

Sandwich Immunosaasy

Brand:  R&D Systems™ 41410-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464234

  • £3312.29 / Pack of 5


Estimated Shipment
26-02-2019
Add to basket

Includes:
  • Pre-coated microtiter plate(s)
  • Plate sealers Wash Solution Concentrate Human Interferon Alpha Standard, 10,000 pg/ml Dilution Buffer Antibody Concentrate HRP Conjugate Concentrate

Description

Description

VeriKine Human IFN-β ELISA kit uses the sandwich immunoassay technique for the quantitative measurement of IFN-β in media. It is developed for superior performance with intra-assay and inter-assay CVs of ≤ 8%.
Specifications

Specifications

Biotin
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates
Human
Sufficient materials to run ELISAs on at least five 96-well plates
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3456
5 Plates
25pg/mL
Cell Culture Supernates (100μL)
Human IFN-β
Videos