missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-beta ELISA Kit, R&D Systems™

Human IFN-beta ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ 41410-1

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454234

  • £814.38 / 96 tests


Estimated Shipment date
from Supplier 27-02-2020
Add to basket

Includes:
  • Pre-coated microtiter plate(s)
  • Plate sealers Wash Solution Concentrate Human Interferon Alpha Standard, 10,000 pg/ml Dilution Buffer Antibody Concentrate HRP Conjugate Concentrate

Description

Description

The Human IFN-beta ELISA Kit quantifies human IFN-beta in cell culture supernates (100 ul).
Specifications

Specifications

1 Kit
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3456
1 Plate
Cell Culture Supernates (100 μL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Cell Culture Supernatant
Biotin
ELISA
Solid Phase Sandwich ELISA
25pg/mL
Human IFN-β
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents