missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-beta ELISA Kit, R&D Systems™
Human IFN-beta ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Target Species | Human |
---|---|
Gene ID (Entrez) | 3456 |
Sample Type | Cell Culture Supernatant |
Sample Volume | Cell Culture Supernates (100 μL) |
Assay Sensitivity | 25pg/mL |
Description
The Human IFN-beta ELISA Kit quantifies human IFN-beta in cell culture supernates (100 ul).Specifications
Human | |
Cell Culture Supernatant | |
25pg/mL |
3456 | |
Cell Culture Supernates (100 μL) |