missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-alpha Serum Sample ELISA Kit, R&D Systems™

Human IFN-alpha Serum Sample ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ 41110-1

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444174

  • £865.83 / 96 tests


Estimated Shipment date
from Supplier 27-03-2020
Add to basket

Description

Description

The Human IFN-alpha Serum Sample ELISA Kit quantifies human IFN-alpha in cell culture supernates (100 ul), serum (100 ul), plasma (100 ul).
Specifications

Specifications

1 Kit
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3440
1 Plate
Cell Culture Supernates (100 μL), Serum (100 μL), Plasma (100 μL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Serum
Biotin
ELISA
Solid Phase Sandwich ELISA
12.5pg/mL
Human IFN-α
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents