missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IFN-alpha Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IFN-alpha.

Brand:  Invitrogen™ BMS216CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571697

  • £372.00 / 96 tests


Estimated Shipment date
from Supplier 05-03-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-alpha
HRP-Conjugate anti-human IFN-alpha monoclonal antibody
Human IFN-alpha Standard, 500pg/mL
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
3.2pg/mL
IFN alpha
96 Tests
Serum
ELISA
20 μL
Alpha interferon, IFN leukocytic, LeIF D
Human
Videos
Documents

For Research Use Only