Learn More
Invitrogen™ HSP90 alpha Human ELISA Kit
For the quantitative detection of human HSP90 alpha.
Brand: Invitrogen™ BMS2090
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP90 alpha
Biotin-Conjugate anti-human HSP90 alpha monoclonal antibody
Streptavidin-HRP
Human HSP90 alpha Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Heat Shock Protein 90 alpha (Hu HSP90 alpha) ELISA quantitates Hu HSP90 alpha in human serum, plasma cell lysate. The assay will exclusively recognize both natural and recombinant Hu HSP90 alpha. Principle of the method The Human HSP90 alpha solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 (MIM 140572), a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).Specifications
P07900 | |
0.03 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
EL52,HSPN,LAP2,HSP86,HSPC1,HSPCA,Hsp89,Hsp90,LAP-2,HSP89A,HSP90A,HSP90N,Hsp103,HSPCAL1,HSPCAL4,HEL-S-65p | |
8.6% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. |
0.156-10 ng/mL | |
Biotin | |
Cell Lysates, Plasma, Serum | |
ELISA | |
3320 | |
10% | |
Pre-coated 96 well plate, Standard, Standard, Sample Diluent, Assay Buffer concentrate, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Controls | |
96 Tests | |
Cell Lysate,100 μL; Plasma, 10 μL; Serum, 10 μL | |
Heat Shock Protein 90 alpha | |
1 hr. 20 min. |