missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit, R&D Systems™

Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DHWAD0

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452665

  • £679.14 / 1 set


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Adiponectin/Acrp30 in serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.989ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Serum
3.90 - 250ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
9370
1 Plate
Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents