missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GM-CSF Quantikine ELISA Kit, R&D Systems™

Human GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SGM00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452776

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 02-04-2020
Add to basket

Description

Description

The Human GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human GM-CSF in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
6 Plates
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Serum
7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
1437
3pg/mL
Natural and recombinant human GM-CSF
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents