missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Follistatin Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Brand:  Gibco™ 120-13-500UG

Additional Details : Weight : 0.16500kg

 View more versions of this product

Product Code. 17831323

  • £2505.00 / 500µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.
Specifications

Specifications

P19883
protein with no preservative
31.5
500 μg
-20°C
<1 EU/μg
Fst
Unconjugated
GNCWLRQAKN GRCQVLYKTE LSKEECCSTG RLSTSWTEED VNDNTLFKWM IFNGGAPNCI PCKETCENVD CGPGKKCRMN KKNKPRCVCA PDCSNITWKG PVCGLDGKTY RNECALLKAR CKEQPELEVQ YQGRCKKTCR DVFCPGSSTC VVDQTNNAYC VTCNRICPEP ASSEQYLCGN DGVTYSSACH LRKATCLLGR SIGLAYEGKC IKAKSCEDIQ CTGGKKCLWD FKVGRGRCSL CDELCPDSKS DEPVCASDNA TYASECAMKE AACSSGVLLE VKHSGSCN
≥ 95% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay, Western Blot
10468
Human Follistatin
E. coli
RUO
activin-binding protein; AL033346; FOL1; Follictatin; Follistatin; follistatin isoform FST317; follistatin-A; FS; FST; Fst-288; RATFOL1
Human
Recombinant
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers