Learn More
R&D Systems™ Human FGF basic Quantikine HS ELISA Kit, R&D Systems™
a 5.5 hour solid phase ELISA designed to measure FGF basic levels in serum, plasma, and urine.
Brand:  R&D Systems™ SSFB00D
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462876
  • £3681.94 / Pack of 6
Estimated Shipment
28-02-2019
Add to basket
Description
Description
It contains E. coli-expressed recombinant human FGF basic and antibodies raised against the recombinant factor. It has been shown to quantitate recombinant human FGF basic accurately. Results obtained using natural human FGF basic showed linear curves that were parallel to the standard curves obtained using the Quantikine HS kit standards. These results indicate that this kit can be used to determine relative mass values for natural human FGF basic
Specifications
Specifications
0.31 - 20pg/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
2247
6 Plates
0.07pg/mL
Serum (100μL), EDTA Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Serum (100uL), EDTA Plasma (100uL), Citrate Plasma (100uL), Urine (100uL)