missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc Receptor Binding Inhibitor Polyclonal Antibody, Functional Grade, eBioscience™, Invitrogen™
Polyclonal Antibody
Brand: Affymetrix eBioscience 16-9161-73
175.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Four different classes of Fcy receptors have been identified: FcyRI (CD64), FcyRII (CD32), and FcyRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcyR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcyR varies depending on the isotype of the monoclonal antibody. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcyRs.Specifications
Fc Receptor Binding Inhibitor | |
20 μL/Test | |
PBS with no preservative; pH 7.2 | |
Human | |
100 Tests | |
Primary | |
4° C |
Control, Flow Cytometry, Functional Assay | |
Functional Grade | |
Fc blocking; FcR BI; FcR Inhibitor | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
For Research Use Only