missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Fc epsilon RI Platinum ELISA Kit

For the detection of human Fc epsilon RI

£357.00 - £2380.00

Specifications

For Use With (Application) ELISA
Sample Size Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
Synonym FCER1, FceRI, Fc epsilon RI
Target Fc epsilon Receptor 1 alpha (FceRI)
Sensitivity 0.068ng/mL
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15581657
View Documents
Invitrogen™
BMS2101
96 Tests £357.00
96 tests View alternative products
Save Up to 
»
Estimated Shipment
04-11-2020
Add to basket
 
15591657
View Documents
Invitrogen™
BMS2101TEN
10 x 96 Tests £2380.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
04-11-2020
Add to basket
 
Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
FCER1, FceRI, Fc epsilon RI
0.068ng/mL
Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
Fc epsilon Receptor 1 alpha (FceRI)
Human