missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Fc epsilon RI Platinum ELISA Kit

For the detection of human Fc epsilon RI

Brand:  Invitrogen™ BMS2101

285.60 GBP valid until 2020-12-18
Use promo code "14718" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581657

  • £357.00 / 96 tests


Estimated Shipment
03-11-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Fc epsilon RI
Detection antibody (chimeric IgE)
HRP-Conjugate anti-human IgE polyclonal antibody
Human Fc epsilon RI Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 1x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
FCER1, FceRI, Fc epsilon RI
Human
0.068ng/mL
Fc epsilon Receptor 1 alpha (FceRI)
96 Tests
Videos
Documents