Learn More
Abnova™ Human FBXW12 Partial ORF (NP_996985.1, 265 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00285231-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
Members of the F-box protein family, such as FBXW12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603034), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: LLRPSEGSDPLSTFLPHKLCASACWTPKVKNRITLMSQSSTGKKTEFITFDLTTKKTGGQTVIQAYEIASFQVAAHLKCPIWMGASDGYMSpecifications
NP_996985.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLRPSEGSDPLSTFLPHKLCASACWTPKVKNRITLMSQSSTGKKTEFITFDLTTKKTGGQTVIQAYEIASFQVAAHLKCPIWMGASDGYM | |
RUO | |
FBXW12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
285231 | |
FBXW12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBXO12/FBXO35/Fbw12/MGC120385/MGC120386/MGC120387 | |
FBXW12 | |
Recombinant | |
wheat germ expression system |