missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ERG Partial ORF (NP_004440.1, 1 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002078-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16003975

  • £275.34 / 10µg

Estimated Shipment
07-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MIQTVPDPAAHIKEALSVVSEDQSLFECAYGTPHLAKTEMTASSSSDYGQTSKMSPRVPQQDWLSQPPARVTIKMECNPSQVNGSRNSPDECSVAKGGKMVGSPDTVGMNYGSYMEEKHM
Specifications

Specifications

NP_004440.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.83kDa
12.5% SDS-PAGE Stained with Coomassie Blue
MIQTVPDPAAHIKEALSVVSEDQSLFECAYGTPHLAKTEMTASSSSDYGQTSKMSPRVPQQDWLSQPPARVTIKMECNPSQVNGSRNSPDECSVAKGGKMVGSPDTVGMNYGSYMEEKHM
RUO
ERG
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
2078
ERG (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
erg-3|p55
ERG
Yes
Videos
Documents