missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Erythropoietin Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ BMS2035TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15561537

  • £2250.00 / Pack of 10


Estimated Shipment date
from Supplier 25-02-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Erythropoietin
Biotin-Conjugate anti-human Erythropoietin monoclonal antibody
Streptavidin-HRP
Human Erythropoietin Standard lyophilized, 200 mIU/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
Specifications

Specifications

P01588
Biotin
2056
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution
RUO
50 μL
2°C to 8°C
1 hr. 25 min.
EPO
6.2%
HRP
Human
1.6-100 mIU/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
0.17 mIU/mL
Plasma (50 μL), Serum (50 μL), Supernatants (50 μL)
1 hr. 20 min.
Human
Erythropoietin, EPO
3.4%
10 x 96 Tests
Supernatant
Videos
Documents

For Research Use Only