Learn More
Invitrogen™ eBioscience™ Human EPO Platinum ELISA Kit, short incubation, for in vitro diagnostic use
For the quantitative detection of human Erythropoietin. Cell culture supernatant, serum and plasma (EDTA, citrate, heparin), were tested with this assay. Erythropoietin (EPO) is a hormone produced by the kidney that promotes the formation of red blood cells in the bone marrow.
Brand: Invitrogen™ BMS2035CE
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human Erythropoietin
Biotin-Conjugate anti-human Erythropoietin monoclonal antibody
Streptavidin-HRP
Human Erythropoietin Standard lyophilized, 200 mIU/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
0.17 mIU/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
Erythropoietin, EPO |
Biotin | |
Human | |
96 Tests | |
Erythropoietin (EPO) |
For Research Use Only