Learn More
Abnova™ Human DHX8 Partial ORF (NP_004932, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001659-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is highly homologous to yeast Prp22. This protein facilitates nuclear export of spliced mRNA by releasing the RNA from the spliceosome. [provided by RefSeq]
Sequence: RREGRVANVADVVSKGQRVKVKVLSFTGTKTSLSMKDVDQETGEDLNPNRRRNLVGETNEETSMRNPDRPTHLSLVSAPEVEDDSLERKRLTRISDPEKWSpecifications
NP_004932 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RREGRVANVADVVSKGQRVKVKVLSFTGTKTSLSMKDVDQETGEDLNPNRRRNLVGETNEETSMRNPDRPTHLSLVSAPEVEDDSLERKRLTRISDPEKW | |
RUO | |
DHX8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1659 | |
DHX8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX8/HRH1/PRP22/PRPF22 | |
DHX8 | |
Recombinant | |
wheat germ expression system |