missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Cytochrome c Quantikine ELISA Kit, R&D Systems™

Human Cytochrome c Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SCTC0

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452696

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 30-03-2020
Add to basket

Description

Description

The Human Cytochrome c Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cytochrome c in cell lysates (20 ul), subcellular fractions (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.31ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Subcellular Fractions
0.63 - 20ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
54205
6 Plates
Cell Lysates (20uL),Subcellular Fractions (100uL)
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents