missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL8/IL-8 DuoSet ELISA, R&D Systems™

Human IL-8/CXCL8 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DY208

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13443235

  • £434.22 / Pack of 15

Estimated Shipment
05-07-2019

Please to purchase this item. Need a web account? Register with us today!


Requires : DuoSet Ancillary Reagent Kit 2 - Catalog # DY008

OR


Wash Buffer - 0.05%Tween™ 20 in PBS, pH 7.2-7.4
Block Buffer - 1% BSA in PBS, pH 7.2-7.4
Reagent Diluent - 0.1% BSA, 0.05% Tween 20 in Tris-buffered Saline, pH 7.2-7.4, 0.2μm filtered
Substrate Solution - Catalog # DY999
Stop Solution - 2N H2SO4
Microplates - Catalog # DY990
ELISA Plate Sealers

Description

Description

The Human IL-8/CXCL8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Capture Antibody
Detection Antibody
Recombinant Standard
Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
1 Kit
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
3576
Solid Phase Sandwich ELISA Development Kit
31.20 to 2,000pg/mL
15 Plates
31.20 - 2000pg/mL
Videos