missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCL12/SDF-1 alpha Quantikine ELISA Kit, R&D Systems™
Human CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 156.00 - 10000pg/mL |
---|---|
Assay Sensitivity | 47pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma (Heparin) |
Description
The Human CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL12/SDF-1 alpha in cell culture supernates (10 ul), edta plasma (100 ul), platelet-poor heparin plasma (100 ul).Specifications
156.00 - 10000pg/mL | |
47pg/mL | |
Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma (Heparin) |