missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL12/SDF-1 alpha Quantikine ELISA Kit, R&D Systems™

Human CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SSA00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452876

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 30-03-2020
Add to basket

Description

Description

The Human CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL12/SDF-1 alpha in cell culture supernates (10 ul), edta plasma (100 ul), platelet-poor heparin plasma (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
Solid Phase Sandwich ELISA
47pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Platelet-poor Plasma (Heparin)
156.00 - 10000pg/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
6387
6 Plates
Cell Culture Supernates (10uL),EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL)
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents