missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL1/GRO alpha Quantikine ELISA Kit, R&D Systems™

Brand:  R&D Systems™ SGR00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462776

  • £3519.18 / Pack of 6


Estimated Shipment date
from Supplier 26-03-2020
Add to basket

Description

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 10pg/mL
Specificity: Natural and recombinant human GRO alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

1 Pack
Human
2919
6 Plates
Cell Culture Supernates (200 μL), Serum (200 μL), EDTA Plasma (200 μL), Heparin Plasma (200 μL), Citrate Plasma (200 μL)
Serum
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Solid Phase Sandwich ELISA
10pg/mL
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents