missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Cu/ZnSOD Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human Cu/ZnSOD.

Brand:  Invitrogen™ BMS222TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571737

  • £2180.00 / Pack of 10


Estimated Shipment date
from Supplier 25-02-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cu/ZnSOD
HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
Human Cu/ZnSOD Standard, 5ng/mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Phosphate Buffered Saline Concentrate (PBS) 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
0.04ng/mL
Cu/ZnSOD
10 x 96 Tests
Urine
ELISA
10 μL (1:20 prediluted)
Cu/Zn Superoxide Dismutase
Human
Videos
Documents

For Research Use Only