missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CTNS Partial ORF (NP_004928, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001497-Q01.25ug
This item is not returnable.
View return policy
Description
This gene encodes a seven-transmembrane domain protein that functions to transport cystine out of lysosomes. Its activity is driven by the H+ electrochemical gradient of the lysosomal membrane. Mutations in this gene cause cystinosis, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MIRNWLTIFILFPLKLVEKCESSVSLTVPPVVKLENGSSTNVSLTLRPPLNATLVITFEITFRSKNITILELPDEVVVPPGVTNSSFQVTSQNVGQLTVYSpecifications
NP_004928 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIRNWLTIFILFPLKLVEKCESSVSLTVPPVVKLENGSSTNVSLTLRPPLNATLVITFEITFRSKNITILELPDEVVVPPGVTNSSFQVTSQNVGQLTVY | |
RUO | |
CTNS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1497 | |
CTNS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTNS-LSB/PQLC4 | |
CTNS | |
Recombinant | |
wheat germ expression system |