missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Clusterin Quantikine ELISA Kit, R&D Systems™

Human Clusterin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DCLU00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452615

  • £621.81 / 96 tests


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

The Human Clusterin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Clusterin/APOJ in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (25 ul), urine (13 ul).
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
1.05ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Urine
3.12 - 200ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
1191
1 Plate
Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (25uL),Urine (13uL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents