Learn More
Invitrogen™ CKMB Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHCKMB
Code : NEW
Additional Details : Weight : 0.00400kg
Description
The Human CKMB ELISA quantitates Hu CKMB in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CKMB. Principle of the method The Human CKMB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Reversibly catalyzes the transfer of phosphate between ATP and various phosphogens (e.g. creatine phosphate). Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.Specifications
P06732,P12277 | |
6 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
1152, 1158 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Cell Cycle & Proliferation | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.328-80 ng/mL | |
0.3ng/mL to 80ng/mL | |
Biotin | |
Kinase & Inhibitors | |
This ELISA pair antibody detects human CKMB. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
B-CK,BCKB,HEL-211,HEL-S-29,CKB,CKMM,M-CK | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
N/A | |
1 hr. 20 min. |