missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHTOP Full-length ORF (NP_056422.2, 1 a.a. - 248 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026097-P01.2ug
This item is not returnable.
View return policy
Description
Sequence: MAAQSAPKVVLKSTTKMSLNERFTNMLKNKQPTPVNIRASMQQQQQLASARNRRLAQQMENRPSVQAALKLKQSLKQRLGKSNIQARLGRPIGALARGAIGGRGLPIIQRGLPRGGLRGGRATRTLLRGGMSLRGQNLLRGGRAVAPRMGLRRGGVRGRGGPGRGGLGRGAMGRGGIGGRGRGMIGRGRGGFGGRGRGRGRGRGALARPVLTKEQLDNQLDAYMSKTKGHLDAELDAYMAQTDPETNDSpecifications
NP_056422.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HT031/C1orf77/FL-SRAG/FOP/RP1-178F15.2/SRAG/SRAG-3/SRAG-5/pp7704 | |
CHTOP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26097 | |
CHTOP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAQSAPKVVLKSTTKMSLNERFTNMLKNKQPTPVNIRASMQQQQQLASARNRRLAQQMENRPSVQAALKLKQSLKQRLGKSNIQARLGRPIGALARGAIGGRGLPIIQRGLPRGGLRGGRATRTLLRGGMSLRGQNLLRGGRAVAPRMGLRRGGVRGRGGPGRGGLGRGAMGRGGIGGRGRGMIGRGRGGFGGRGRGRGRGRGALARPVLTKEQLDNQLDAYMSKTKGHLDAELDAYMAQTDPETND | |
RUO | |
CHTOP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |