missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human CD258 (LIGHT) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of protein levels from samples including serum, plasma, and cell culture supernatants. CD258, or LIGHT, is a 29 kd type II transmembrane protein and a member of the TNF ligand superfamily.

Brand:  Invitrogen™ 88-7258-86

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15521087

  • £292.87 / Pack of 10


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

Description

Description

Human CD258 (LIGHT) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
10 x 96 tests
Human
CD258 (LIGHT)
96-well plates
75pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
TNFSF14, TR2, HVEML, LTg
Videos

For Research Use Only