missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL5/RANTES Quantikine ELISA Kit, R&D Systems™

Human CCL5/RANTES Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SRN00B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432866

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 02-03-2020
Add to basket

Description

Description

The Human CCL5/RANTES Quantikine ELISA Kit quantifies human CCL5/RANTES in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
6 Plates
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL)
Sufficient materials to run ELISAs on at least six 96-well plates
Urine
31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6352
6.6pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
Documents