Learn More
Invitrogen™ CCL3 (MIP-1 alpha) Human Uncoated ELISA Kit with Plates
Uncoated ELISA Kit with Plates
Brand: Invitrogen™ 88-7035-22
110.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.47500kg
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
The Human CCL3 (MIP-1 alpha) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human CCL3 (MIP-1 alpha). Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
CCL3 (MIP1a, Macrophage Inflammatory Protein-1 alpha, MIP-1 alpha) has a molecular weight of approximately 8 kDa, belongs to the family of chemotactic cytokines, and is involved in chemotactic and proinflammatory effects, and homeostasis. CCL3 is produced by lymphocytes, macrophages and dendritic cells, and is involved in inflammatory response of blood monocytes and tissue macrophages. Also, CCL3 can exist as a naturally occurring heterodimer with MIP-1beta and has been shown to have antiviral activity against HSV-1. CCL3 protein can bind with high affinity to the CCR1 and CCR5 receptors.Specifications
P10147 | |
16 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
BCGF-1,BCGF1,BSF-1,BSF1,IL-4 | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
16-2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
6348 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Streptavidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
2 x 96 Tests | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
C-C motif chemokine 3, MIP1A, Macrophage 1nflammatory Protein-1 alpha | |
1 hr. 45 min. |
For Research Use Only