Learn More
Invitrogen™ MCP-1/CCL2 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human CCL2 protein levels from samples including serum, plasma, and cell culture supernatants.
Brand: Invitrogen™ 88-7399-77
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
MCP-1 (Monocyte chemotactic protein-1, CCL2) has a predicted molecular weight of approximately 11kDa, however, it may run at a higher molecular weight due to glycosylation. MCP-1 is a member of the cytokine family and plays an important role in immunoregulation and inflammatory processes. MCP-1 signals through G protein-coupled receptors, CCR2 and CCR4, to recruit memory T cells, monocytes and dendritic cells. MCP-1 is linked with tumor progression, angiogenesis, and displays selective chemotactic activity towards monocytes and basophils but not neutrophils or eosinophils. MCP-1 is produced by a variety of cell types and is a potent chemoattractant for monocytes, memory T lymphocytes, and natural killer (NK) cells. Further, MCP-1 is indicated in the etiology of several immune disorders, and is upregulated during infection and inflammation. While MCP-1 is a potent basophil activator, the related protein MCP-2 stimulates both eosinophils and basophils. MCP-3 has been shown to have the broadest range of influence. MCP-1 has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis or atherosclerosis.Specifications
P13500 | |
7 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IFN-g,Ifg | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
7-1,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
6347 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Streptavidin-HRP Concentrate (100X), Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
20 x 96 Tests | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Monocyte chemotactic protein 1 | |
1 hr. 45 min. |
For Research Use Only