missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL17/TARC Quantikine ELISA Kit
Human CCL17/TARC Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 31.20 to 2000 pg/mL |
---|---|
Assay Sensitivity | 7 pg/mL |
Sample Type | Cell Culture Supernatants, Serum, Plasma (EDTA, Heparin), Plasma (EDTA), Plasma (Heparin), Serum |
Description
The Human CCL17/TARC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL17/TARC in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Specifications
31.20 to 2000 pg/mL | |
7 pg/mL | |
Cell Culture Supernatants, Serum, Plasma (EDTA, Heparin), Plasma (EDTA), Plasma (Heparin), Serum |