missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL17/TARC Quantikine ELISA Kit, R&D Systems™

Human CCL17/TARC Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SDN00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462696

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 30-03-2020
Add to basket

Description

Description

The Human CCL17/TARC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL17/TARC in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
Solid Phase Sandwich ELISA
7pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Serum
31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6361
6 Plates
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents