Learn More
Invitrogen™ Cathepsin L Human ELISA Kit
For the quantitative detection of human Cathepsin L.
Brand: Invitrogen™ BMS257TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cathepsin L
Biotin-Conjugate anti-human Cathepsin L monoclonal antibody
Streptavidin-HRP
Human Cathepsin L Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Cathepsin L ELISA quantitates Hu Cathepsin L in human serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Cathepsin L. Principle of the method The Human Cathepsin L solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene.Specifications
P07711 | |
1.7 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
CATL1,MEP,CTSL | |
6.2% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
3.1-50 ng/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
1514 | |
6% | |
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Assay Buffer concentrate, Biotinylated Detection Antibody, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Lysis Buffer, Stop Solution, Stop Solution | |
10 x 96 Tests | |
Serum, 50 μL; Supernatant, 50 μL | |
Major excreted protein, MEP, Cathepsin L precursor | |
1 hr. 20 min. |
For Research Use Only